This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00513948_g1
See other MIB2 GE Assays ›
Gene Symbol:
Mib2
Gene Name:
mindbomb homolog 2 (Drosophila)
Gene Aliases:
2210008I11Rik, Zzank1, skd
Chromosome Location:
Chr.4: 155654470 - 155669254 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00513948_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mib2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256107.1 NP_001243036.1 5 - 6 1384 61
NM_001256108.2 NP_001243037.2 6 - 7 1300 61
NM_145124.3 NP_660106.2 5 - 6 1384 61
GenBank mRNA AB063290.1 - 3 - 4 389 61
AB072336.1 - 5 - 6 946 61
AK147940.1 - 6 - 7 1244 61
AK159611.1 - 5 - 6 1102 61
AY974090.1 - 3 - 4 525 61
BC058086.1 - 4 - 5 643 61
BC141133.1 - 4 - 5 698 9055745 61
BC145441.1 - 4 - 5 698 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400004147.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mib2

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
Notch signaling pathway
protein ubiquitination
positive regulation of I-kappaB kinase/NF-kappaB signaling
actin binding
ubiquitin-protein transferase activity
signal transducer activity
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400034797.mm.1 NM_001256107.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034797.mm.1 NM_001256107.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034797.mm.1 NM_001256108.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400034797.mm.1 NM_145124.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034797.mm.1 NM_145124.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400034797.mm.1 TC0400004147.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline