This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00514143_g1
See other GTF3C5 GE Assays ›
Gene Symbol:
Gtf3c5
Gene Name:
general transcription factor IIIC, polypeptide 5
Gene Aliases:
2700084A09Rik, TFIIIC63, TFiiiC2-63
Chromosome Location:
Chr.2: 28566245 - 28583746 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00514143_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf3c5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290484.1 NP_001277413.1 6 - 7 1295 96
NM_148928.3 NP_683730.1 6 - 7 1295 96
GenBank mRNA AK045419.1 - 6 - 7 887 96
AK147014.1 - 6 - 7 1273 96
BC027055.1 - 6 - 7 1262 5360397 96
BC027247.1 - 6 - 7 1205 2631223 96
BC039171.1 - 6 - 7 1216 4500799 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003233.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf3c5

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
skeletal muscle cell differentiation
molecular_function
DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200027258.mm.1 NM_148928.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200027258.mm.1 TC0200003233.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0