This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00514641_g1
See other ECM1 GE Assays ›
Gene Symbol:
Ecm1
Gene Name:
extracellular matrix protein 1
Gene Aliases:
AI663821, p85
Chromosome Location:
Chr.3: 95734147 - 95739819 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00514641_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ecm1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252653.1 NP_001239582.1 8 - 9 1269 60
NM_007899.2 NP_031925.2 8 - 9 1272 60
GenBank mRNA AK153009.1 - 8 - 9 1271 60
AK153471.1 - 8 - 9 1271 60
AK159334.1 - 8 - 9 1268 60
AK169939.1 - 8 - 9 1274 60
BC138693.1 - 8 - 9 1258 8861715 60
BC145381.1 - 8 - 9 1255 9053939 60
L33416.1 - 8 - 9 1265 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002482.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ecm1

Gene Ontology Categories:

Function(s) Process(es)

ossification
angiogenesis
positive regulation of endothelial cell proliferation
negative regulation of cytokine-mediated signaling pathway
regulation of type 2 immune response
regulation of transcription from RNA polymerase II promoter
inflammatory response
signal transduction
negative regulation of peptidase activity
regulation of bone mineralization
negative regulation of bone mineralization
biomineral tissue development
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of angiogenesis
regulation of T cell migration
protease binding
signal transducer activity
interleukin-2 receptor binding
protein C-terminus binding
enzyme binding
laminin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300019248.mm.1 NM_001252653.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300019248.mm.1 NM_007899.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300019248.mm.1 TC0300002482.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0