This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00515106_m1
See other HDAC2 GE Assays ›
Gene Symbol:
Hdac2
Gene Name:
histone deacetylase 2
Gene Aliases:
D10Wsu179e, YAF1, Yy1bp, mRPD3
Chromosome Location:
Chr.10: 36974544 - 37001889 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00515106_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hdac2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008229.2 NP_032255.2 2 - 3 373 90
GenBank mRNA AB221663.1 - 2 - 3 165 90
AK051753.1 - 2 - 3 348 90
BC138517.1 - 2 - 3 345 8861537 90
U31758.1 - 2 - 3 333 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000312.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hdac2

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
maintenance of chromatin silencing
transcription, DNA-templated
regulation of transcription, DNA-templated
protein deacetylation
positive regulation of cell proliferation
epidermal cell differentiation
positive regulation of epithelial to mesenchymal transition
positive regulation of receptor biosynthetic process
negative regulation of neuron projection development
dendrite development
chromatin modification
histone deacetylation
hippocampus development
neuron differentiation
positive regulation of interleukin-1 production
positive regulation of tumor necrosis factor production
circadian regulation of gene expression
odontogenesis of dentin-containing tooth
positive regulation of tyrosine phosphorylation of Stat3 protein
embryonic digit morphogenesis
ATP-dependent chromatin remodeling
negative regulation of apoptotic process
negative regulation of DNA binding
negative regulation of sequence-specific DNA binding transcription factor activity
positive regulation of proteolysis
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
rhythmic process
positive regulation of oligodendrocyte differentiation
positive regulation of sequence-specific DNA binding transcription factor activity
regulation of protein kinase B signaling
cardiac muscle cell development
negative regulation of cardiac muscle cell proliferation
regulation of sarcomere organization
hair follicle placode formation
negative regulation of dendritic spine development
eyelid development in camera-type eye
fungiform papilla formation
cellular response to hydrogen peroxide
histone H3 deacetylation
histone H4 deacetylation
negative regulation of canonical Wnt signaling pathway
regulation of protein deacetylation
negative regulation of peptidyl-lysine acetylation
negative regulation of intrinsic apoptotic signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
core promoter binding
RNA polymerase II repressing transcription factor binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
histone deacetylase activity
protein binding
transcription factor binding
hydrolase activity
deacetylase activity
enzyme binding
heat shock protein binding
histone deacetylase activity (H3-K14 specific)
chromatin DNA binding
nucleosomal DNA binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
histone deacetylase activity (H3-K9 specific)
protein deacetylase activity
histone deacetylase activity (H4-K16 specific)
Krueppel-associated box domain binding
sequence-specific DNA binding
poly(A) RNA binding
NAD-dependent histone deacetylase activity (H3-K9 specific)
NAD-dependent histone deacetylase activity (H4-K16 specific)
NF-kappaB binding
NAD-dependent histone deacetylase activity (H3-K18 specific)

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000002048.mm.1 NM_008229.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000002048.mm.1 NM_008229.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000002048.mm.1 TC1000000312.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0