This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00515458_m1
See other CDH11 GE Assays ›
Gene Symbol:
Cdh11
Gene Name:
cadherin 11
Gene Aliases:
Cad11
Chromosome Location:
Chr.8: 102632095 - 102785480 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00515458_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009866.4 NP_033996.4 2 - 3 330 70
GenBank mRNA AK045023.1 - 2 - 3 299 70
AK049652.1 - 2 - 3 296 70
BC046314.1 - 2 - 3 230 6312939 70
CB246151.1 - 2 - 3 573 70
D21253.1 - 1 - 2 93 70
D31963.1 - 2 - 3 275 70
X77557.1 - 2 - 3 239 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002783.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK049652.1,BC046314.1,D21253.1,D31963.1,NM_009866.4,X77557.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdh11

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
corticospinal tract morphogenesis
calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800021332.mm.1 NM_009866.5 Refseq
Affymetrix Mouse Clariom S Array PSR0800021332.mm.1 TC0800002783.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800021333.mm.1 NM_009866.5 Refseq
Affymetrix Mouse Clariom S Array PSR0800021333.mm.1 TC0800002783.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline