This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00515462_m1
See other CDH11 GE Assays ›
Gene Symbol:
Cdh11
Gene Name:
cadherin 11
Gene Aliases:
Cad11
Chromosome Location:
Chr.8: 102632095 - 102785480 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00515462_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009866.4 NP_033996.4 6 - 7 1317 101
GenBank mRNA AK028271.1 - 2 - 3 204 101
AK045023.1 - 6 - 7 1286 101
AK049652.1 - 6 - 7 1283 101
BC046314.1 - 6 - 7 1217 6312939 101
D21253.1 - 5 - 6 1080 101
D31963.1 - 6 - 7 1262 101
X77557.1 - 6 - 7 1226 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002783.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdh11

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
corticospinal tract morphogenesis
calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800021331.mm.1 NM_009866.5 Refseq
Affymetrix Mouse Clariom S Array PSR0800021331.mm.1 TC0800002783.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800021329.mm.1 NM_009866.5 Refseq
Affymetrix Mouse Clariom S Array PSR0800021329.mm.1 TC0800002783.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800021330.mm.1 NM_009866.5 Refseq
Affymetrix Mouse Clariom S Array PSR0800021330.mm.1 TC0800002783.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 923718 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline