This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00515913_g1
See other HDAC3 GE Assays ›
Gene Symbol:
Hdac3
Gene Name:
histone deacetylase 3
Gene Aliases:
AW537363
Chromosome Location:
Chr.18: 37936971 - 37955000 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00515913_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hdac3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010411.2 NP_034541.2 11 - 12 997 76
GenBank mRNA AF074881.1 - 11 - 12 944 76
AF074882.1 - 5 - 6 371 76
AF098295.1 - 11 - 12 922 76
AF125536.1 - 11 - 12 929 76
AK037478.1 - 472 76
AK145158.1 - 11 - 12 963 76
BC139300.1 - 11 - 12 1025 8862322 76
BC139301.1 - 11 - 12 1025 8862323 76
JN651901.1 - 6 - 7 396 76
JN651902.1 - 5 - 6 354 76
JN651903.1 - 5 - 6 354 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001220.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hdac3

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
positive regulation of protein phosphorylation
transcription, DNA-templated
regulation of transcription, DNA-templated
protein deacetylation
regulation of mitotic cell cycle
chromatin modification
histone deacetylation
regulation of protein stability
positive regulation of TOR signaling
negative regulation of interleukin-1 production
negative regulation of tumor necrosis factor production
circadian regulation of gene expression
regulation of multicellular organism growth
positive regulation of transcription factor import into nucleus
negative regulation of sequence-specific DNA binding transcription factor activity
positive regulation of neuron apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of JNK cascade
negative regulation of protein export from nucleus
spindle assembly
cellular response to fluid shear stress
positive regulation of type B pancreatic cell apoptotic process
negative regulation of cardiac muscle cell differentiation
DNA binding
chromatin binding
transcription corepressor activity
histone deacetylase activity
protein binding
transcription factor binding
hydrolase activity
enzyme binding
cyclin binding
histone deacetylase activity (H3-K14 specific)
chromatin DNA binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
histone deacetylase activity (H3-K9 specific)
protein deacetylase activity
histone deacetylase activity (H4-K16 specific)
histone deacetylase binding
NAD-dependent histone deacetylase activity (H3-K9 specific)
NAD-dependent histone deacetylase activity (H4-K16 specific)
GTPase binding
NF-kappaB binding
NAD-dependent histone deacetylase activity (H3-K18 specific)

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800008982.mm.1 NM_010411.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800008982.mm.1 NM_010411.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800008982.mm.1 TC1800001220.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0