This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00516016_m1
See other ELAVL2 GE Assays ›
Gene Symbol:
Elavl2
Gene Name:
ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B)
Gene Aliases:
Hub, mel-N1
Chromosome Location:
Chr.4: 91250766 - 91400064 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00516016_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Elavl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010486.2 NP_034616.1 5 - 6 885 99
GenBank mRNA AY035380.1 - 4 - 5 734 99
U29088.1 - 5 - 6 884 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003043.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Elavl2

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Process(es)

nucleotide binding
nucleic acid binding
RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400024439.mm.1 TC0400003043.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400024439.mm.1 NM_001177883.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400024439.mm.1 NM_001177883.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400024439.mm.1 NM_010486.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400024439.mm.1 NM_010486.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400024439.mm.1 NM_207685.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400024439.mm.1 NM_207685.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0