Assay Details
| Gene Symbol: Il16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010551.3 | NP_034681.2 | 16 - 17 | 3589 | 90 | |
| GenBank mRNA | AF006001.1 | - | 3 - 4 | 1319 | 90 | |
| AF017111.1 | - | 4 - 5 | 1469 | 90 | ||
| AF175292.1 | - | 16 - 17 | 3589 | 90 | ||
| AK038319.1 | - | 4 - 5 | 1505 | 90 | ||
| AK169551.1 | - | 4 - 5 | 1492 | 90 | ||
| AK220455.1 | - | 16 - 17 | 3599 | 90 | ||
| BC026894.1 | - | 4 - 5 | 1398 | 90 | ||
| BC058709.1 | - | 16 - 17 | 6841305 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il16
Genomic Map