This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00516259_m1
See other ADGRE5 GE Assays ›
Gene Symbol:
Adgre5
Gene Name:
adhesion G protein-coupled receptor E5
Gene Aliases:
AA409984, Cd97, TM7LN1
Chromosome Location:
Chr.8: 83723250 - 83741311 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00516259_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgre5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163029.1 NP_001156501.1 10 - 11 1302 114
NM_001163030.1 NP_001156502.1 11 - 12 1449 114
NM_001163031.1 NP_001156503.1 10 - 11 1296 114
NM_011925.2 NP_036055.2 12 - 13 1584 114
GenBank mRNA AK004577.1 - 10 - 11 1312 114
BC006676.1 - 10 - 11 1164 3499451 114
CT010326.1 - 10 - 11 1128 114
Y18365.1 - 12 - 13 1442 114

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002558.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adgre5

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
positive regulation of synapse assembly
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800019480.mm.1 NM_001163029.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019480.mm.1 NM_001163029.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019480.mm.1 NM_001163030.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019480.mm.1 NM_001163030.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019480.mm.1 NM_001163031.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019480.mm.1 NM_001163031.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019480.mm.1 NM_011925.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019480.mm.1 NM_011925.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800019480.mm.1 TC0800002558.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800019481.mm.1 NM_001163029.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019481.mm.1 NM_001163029.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019481.mm.1 NM_001163030.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019481.mm.1 NM_001163030.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019481.mm.1 NM_001163031.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019481.mm.1 NM_001163031.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019481.mm.1 NM_011925.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019481.mm.1 NM_011925.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800019481.mm.1 TC0800002558.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800019479.mm.1 NM_001163029.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019479.mm.1 NM_001163029.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019479.mm.1 NM_001163030.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019479.mm.1 NM_001163030.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019479.mm.1 NM_001163031.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019479.mm.1 NM_001163031.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019479.mm.1 NM_011925.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019479.mm.1 NM_011925.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800019479.mm.1 TC0800002558.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800019482.mm.1 NM_001163029.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019482.mm.1 NM_001163029.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019482.mm.1 NM_001163030.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019482.mm.1 NM_001163030.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019482.mm.1 NM_001163031.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019482.mm.1 NM_001163031.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019482.mm.1 NM_011925.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019482.mm.1 NM_011925.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800019482.mm.1 TC0800002558.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4d4pq:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0