This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00516995_m1
See other EEF1B2 GE Assays ›
Gene Symbol:
Eef1b2
Gene Name:
eukaryotic translation elongation factor 1 beta 2
Gene Aliases:
2810017J07Rik, Eef1b
Chromosome Location:
Chr.1: 63176831 - 63180486 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00516995_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eef1b2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018796.3 NP_061266.2 1 - 2 464 55
GenBank mRNA AF029844.2 - 1 - 2 130 55
AK012756.1 - 1 - 2 167 55
AK027911.1 - 1 - 2 167 55
AK168191.1 - 1 - 2 166 55
BC003899.1 - 1 - 2 160 3600850 55
BC023139.1 - 1 - 2 176 3963930 55
BC039635.1 - 1 - 2 179 55

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000519.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eef1b2

Panther Classification:

Molecular Function -

translation elongation factor

Gene Ontology Categories:

Function(s) Process(es)

translation
translational elongation
translation elongation factor activity
protein binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20285515 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20285517 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20285516 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P412595 AF029844.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P412595 AF029844.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 36701 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 315955 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177185 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P412595 AF029844.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P412595 AF029844.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P412595 AF029844.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P412595 AF029844.2 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20285528 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20285517 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20285526 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20285521 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20285544 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20285536 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20285523 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20285541 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20285537 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20285524 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20285519 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20285522 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20285538 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20285527 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20285515 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20285529 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20285534 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177185 - -
Whole Mouse Genome G4122A A_51_P412595 AF029844.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P412595 AF029844.2 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177185 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00488056 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00488054 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20285517 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20285516 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20285515 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20285515 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20285544 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20285528 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20285526 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20285514 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20285517 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20285523 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20285519 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20285522 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20285538 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20285521 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20285512 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20285510 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20285536 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20285534 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20285541 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20285543 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20285526 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00488082 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20285519 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20285534 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20285537 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20285542 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20285541 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20285517 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20285544 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20285514 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20285512 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20285524 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20285543 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20285523 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20285538 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20285515 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20285528 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20285536 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10346868 - -
Affymetrix Mouse Gene 1.0 ST Array 10346871 - -
Affymetrix Mouse Gene 1.0 ST Array 10346869 - -
Affymetrix Mouse Gene 1.0 ST Array 10346872 - -
Affymetrix Mouse Gene 1.0 ST Array 10346870 - -
Affymetrix HT MG-430 PM Array Plate 1448252_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10346867 - -
Mouse Genome 430 2.0 Array 1448252_PM_a_at - -
Mouse Genome 430 2.0 Array 1448252_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 55949_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 55949_PM_at - -
Affymetrix Mouse Clariom S Array PSR0100004156.mm.1 NM_018796.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100004156.mm.1 NM_018796.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100004156.mm.1 TC0100000519.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100004157.mm.1 NM_018796.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100004157.mm.1 NM_018796.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100004157.mm.1 TC0100000519.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100004158.mm.1 NM_018796.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100004158.mm.1 NM_018796.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100004158.mm.1 TC0100000519.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100004159.mm.1 NM_018796.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100004159.mm.1 NM_018796.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100004159.mm.1 TC0100000519.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100004171.mm.1 NM_018796.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100004171.mm.1 NM_018796.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100004171.mm.1 TC0100000519.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100004161.mm.1 NM_018796.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100004161.mm.1 NM_018796.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100004161.mm.1 TC0100000519.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100004152.mm.1 NM_018796.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100004152.mm.1 NM_018796.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100004152.mm.1 TC0100000519.mm.1 MTA TC
Mouse Expression Set 430 1448252_PM_a_at - -
Mouse Expression Set 430 1448252_a_at - -
Murine Genome U74v2 Set 96624_r_at - -
Murine Genome U74v2 Set 96624_PM_r_at - -
Murine Genome U74v2 Set 169018_PM_at - -
Murine Genome U74v2 Set 141174_PM_at - -
Murine Genome U74v2 Set 169018_at - -
Murine Genome U74v2 Set 141174_at - -
Mouse Genome 430A 2.0 Array 1448252_PM_a_at - -
Mouse Genome 430A 2.0 Array 1448252_a_at - -
Affymetrix HT Mouse Genome 430A Array 1448252_a_at - -
Affymetrix HT Mouse Genome 430A Array 1448252_PM_a_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 907395 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 6130161 - -
Illumina mouse-6 v1.0 expression beadchip 4850400 - -
Illumina mouse-6 v1.0 expression beadchip 6130161 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004850400 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006130161 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2708644 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1229042 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2708644 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1229042 - -
mouse-6 v1.1 (Illumina) scl000993.1_5-S - -
mouse-6 v1.1 (Illumina) scl055949.3_17-S - -
Illumina Sentrix mouse-6.v1 bead array 6130161 - -
Illumina Sentrix mouse-6.v1 bead array 4850400 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl000993.1_5-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl055949.3_17-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4850400 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6130161 - -
MouseRef-8 v2 Expression BeadChip ILMN_2708644 - -
MouseRef-8 v2 Expression BeadChip ILMN_1229042 - -
MouseRef-8 v2 Expression BeadChip ILMN_2874853 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2708644 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229042 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2874853 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000993.1_5-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl055949.3_17-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lsr54:80/100.66.79.61:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0