This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00517131_m1
See other BACE2 GE Assays ›
Gene Symbol:
Bace2
Gene Name:
beta-site APP-cleaving enzyme 2
Gene Aliases:
1110059C24Rik, AEPLC, AI850424, ALP56, ARP1, ASP21, BAE2, CDA13, CEAP1, DRAP
Chromosome Location:
Chr.16: 97356728 - 97439012 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00517131_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bace2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019517.4 NP_062390.3 1 - 2 368 71
GenBank mRNA AF216310.1 - 1 - 2 319 71
AK052309.1 - 1 - 2 355 71
AK078770.1 - 1 - 2 335 71
AK165036.1 - 1 - 2 304 71
BC120773.1 - 1 - 2 334 40129696 71
BC131947.1 - 1 - 2 334 40130224 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001089.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bace2

Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
membrane protein ectodomain proteolysis
protein catabolic process
negative regulation of amyloid precursor protein biosynthetic process
beta-amyloid metabolic process
beta-amyloid binding
aspartic-type endopeptidase activity
peptidase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600008619.mm.1 NM_019517.5 Refseq
Affymetrix Mouse Clariom S Array PSR1600008619.mm.1 TC1600001089.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600008617.mm.1 NM_019517.5 Refseq
Affymetrix Mouse Clariom S Array PSR1600008617.mm.1 TC1600001089.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600008618.mm.1 NM_019517.5 Refseq
Affymetrix Mouse Clariom S Array PSR1600008618.mm.1 TC1600001089.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline