This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00517764_m1
See other GNGT2 GE Assays ›
Gene Symbol:
Gngt2
Gene Name:
guanine nucleotide binding protein (G protein), gamma transducing activity polypeptide 2
Gene Aliases:
AV096488
Chromosome Location:
Chr.11: 95842219 - 95845734 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00517764_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gngt2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038664.2 NP_001033753.2 2 - 3 213 61
NM_001284393.1 NP_001271322.1 2 - 3 217 61
GenBank mRNA AA028742.1 - 2 - 3 168 61
AK010554.1 - 1 - 2 1227 61
BC061005.1 - 2 - 3 163 30295552 61
BE847751.1 - 2 - 3 203 61
W35863.1 - 2 - 3 222 61

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK010554.1, BC061005.1, NM_001038664.2, NM_001284393.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
metabolic process
signal transducer activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline