This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00518346_m1
See other ECHDC2 GE Assays ›
Gene Symbol:
Echdc2
Gene Name:
enoyl Coenzyme A hydratase domain containing 2
Gene Aliases:
1300017C12Rik, 2610009M20Rik, D4Ertd765e
Chromosome Location:
Chr.4: 108165436 - 108179308 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00518346_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Echdc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001254754.1 NP_001241683.1 7 - 8 790 71
NM_026728.4 NP_081004.2 8 - 9 858 71
GenBank mRNA AK005030.1 - 8 - 9 799 71
AK011365.1 - 7 - 8 736 71
AK153878.1 - 8 - 9 833 71
AK160482.1 - 6 - 7 561 71
AK166388.1 - 8 - 9 71
AK166965.1 - 8 - 9 800 71
BC025104.1 - 8 - 9 3591648 71
BC025454.1 - 4 - 5 342 5352625 71

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Echdc2

Panther Classification:

Molecular Function -

hydratase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
metabolic process
catalytic activity
lyase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0