Assay Details
| Gene Symbol: Creb3l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB047647.1 | - | 7 - 8 | 756 | 103 | |
| AB052779.1 | - | 9 - 10 | 1013 | 103 | ||
| AB182651.1 | - | 9 - 10 | 1131 | 103 | ||
| AB182652.1 | - | 7 - 8 | 756 | 103 | ||
| AF287260.1 | - | 7 - 8 | 894 | 103 | ||
| AK005918.1 | - | 7 - 8 | 913 | 103 | ||
| AK019928.1 | - | 9 - 10 | 1131 | 103 | ||
| AK088919.1 | - | 9 - 10 | 1030 | 103 | ||
| AY049978.1 | - | 9 - 10 | 1052 | 103 | ||
| BC022605.1 | - | 9 - 10 | 987 | 4164994 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22324141 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22324140 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497882 | BC022605.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497882 | AK019928.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497882 | AK088919.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497882 | AB052779.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497882 | AB182651.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497882 | BC022605.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497882 | AK019928.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497882 | AK088919.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497882 | AB052779.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497882 | AB182651.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921694 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 182481 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 653351 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 25437 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152835 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497882 | BC022605.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497882 | AK019928.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497882 | AK088919.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497882 | AB052779.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497882 | AB182651.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497882 | BC022605.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497882 | AK019928.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497882 | AK088919.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497882 | AB052779.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497882 | AB182651.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497882 | BC022605.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497882 | AK019928.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497882 | AK088919.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497882 | AB052779.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497882 | AB182651.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497882 | BC022605.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497882 | AK019928.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497882 | AK088919.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497882 | AB052779.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P497882 | AB182651.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22324124 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152835 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04985938 | - | - |
| Whole Mouse Genome G4122A | A_51_P497882 | BC022605.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497882 | AK019928.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497882 | AK088919.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497882 | AB052779.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497882 | AB182651.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497882 | BC022605.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497882 | AK019928.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497882 | AK088919.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497882 | AB052779.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P497882 | AB182651.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109665 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152835 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03107461 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03107457 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03107462 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03107472 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02786633 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02989976 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03107450 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02989974 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03107460 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22324141 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22324140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22324157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02823588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22324076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02823584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03107457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22324127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22324075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22324104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22324108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02823579 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02823586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22324101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02838902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22324110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22324072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02823570 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22324096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02823583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22324073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03107478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22324125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22324121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22324132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22324124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02823568 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22324117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03064094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22324071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02453456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03107460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22324103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02823577 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22324099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02786632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03107462 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10499762 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499765 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499756 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499755 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499760 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499753 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499752 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499759 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499758 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499754 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499761 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499763 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499757 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424218_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10499751 | - | - |
| Mouse Genome 430 2.0 Array | 1424218_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1424218_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78284_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78284_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300018356.mm.1 | NM_030080.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018356.mm.1 | TC0300002367.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424218_PM_a_at | - | - |
| Mouse Expression Set 430 | 1424218_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424218_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424218_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424218_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424218_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1980736 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1980736 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001980736 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213549 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213549 | - | - |
| mouse-6 v1.1 (Illumina) | scl21922.11.1_7-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1980736 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21922.11.1_7-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1980736 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1213549 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213549 | - | - |

Creb3l4
Genomic Map