This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00519163_g1
See other CNTNAP4 GE Assays ›
Gene Symbol:
Cntnap4
Gene Name:
contactin associated protein-like 4
Gene Aliases:
Caspr4, E130114F09Rik
Chromosome Location:
Chr.8: 112569965 - 112882707 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00519163_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cntnap4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_130457.2 NP_569724.2 5 - 6 842 104
GenBank mRNA AF333770.1 - 5 - 6 807 104
AK042607.1 - 5 - 6 831 104
AK053611.1 - 5 - 6 834 104
AK081891.1 - 5 - 6 835 104
AK142816.1 - 5 - 6 838 104
AK173255.1 - 5 - 6 817 104
BC034628.1 - 5 - 6 842 5369648 104

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001349.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cntnap4

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
regulation of synaptic transmission, dopaminergic
regulation of synaptic transmission, GABAergic
regulation of grooming behavior
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800011082.mm.1 NM_130457.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011082.mm.1 NM_130457.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800011082.mm.1 TC0800001349.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800011084.mm.1 NM_130457.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011084.mm.1 NM_130457.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800011084.mm.1 TC0800001349.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800011083.mm.1 NM_130457.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011083.mm.1 NM_130457.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800011083.mm.1 TC0800001349.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0