This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00519581_m1
See other PSMG2 GE Assays ›
Gene Symbol:
Psmg2
Gene Name:
proteasome (prosome, macropain) assembly chaperone 2
Gene Aliases:
1700017I17Rik, AW545363, Clast3, Tnfsf5ip1
Chromosome Location:
Chr.18: 67641596 - 67654181 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00519581_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Psmg2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134138.1 NP_598899.1 1 - 2 141 64
GenBank mRNA AB031387.1 - 1 - 2 141 64
AK006069.1 - 1 - 2 142 64
AK077569.1 - 1 - 2 128 64
AK135578.1 - 1 - 2 133 64
BC016606.1 - 1 - 2 120 2646038 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000708.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Psmg2

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

mitotic spindle assembly checkpoint
negative regulation of apoptotic process
proteasome assembly
regulation of cell cycle
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31792975 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31792971 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31792973 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31792968 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31792969 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31792970 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31792976 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31792974 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043009 AK077569.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043009 NM_134138.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043009 AB031387.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 232909 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779362 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 657013 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 98003 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73867 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402218 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 196144 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837617 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 879621 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 36976 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104691 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141663 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043009 AK077569.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043009 NM_134138.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043009 AB031387.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792955 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792944 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792957 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792968 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792960 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792992 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792973 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792953 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792964 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792966 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792990 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792958 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792976 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792942 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792965 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792947 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792989 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792971 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792954 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792941 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792969 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792943 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792975 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792984 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792981 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792979 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792974 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792951 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792962 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792949 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792994 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792986 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792978 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31792963 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104691 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141663 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09037098 - -
Whole Mouse Genome G4122A A_52_P403352 AK135578.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P403352 AK167840.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P403352 AK077569.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P403352 BC016606.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P403352 NM_134138.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P338515 AK167840.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P338515 AK077569.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P338515 NM_134138.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P338515 AB031387.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101387 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106718 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104691 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141663 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13871049 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13871051 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31792976 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31792968 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31792970 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31792975 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31792974 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31792973 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31792971 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31792969 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792973 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793021 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792994 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792984 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793062 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793066 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793033 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793030 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793071 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793034 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793014 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793059 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792978 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792998 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793041 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793017 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793019 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792975 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793057 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792981 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793029 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793065 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793015 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793052 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793047 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792992 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793004 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793072 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793000 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793045 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792987 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792990 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793012 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793032 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793010 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793068 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792976 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792996 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792974 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793039 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793040 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793026 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793025 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793036 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793042 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793009 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31793056 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31792979 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31792976 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793025 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793024 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31792998 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793050 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793023 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793002 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793049 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31792981 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793041 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14071313 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13871049 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793019 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793004 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13957654 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13923042 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793051 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13994212 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31792992 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14029131 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31792994 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13648838 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31792999 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31792982 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793012 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793015 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793010 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31792990 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793026 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793014 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793006 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793047 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31792996 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793017 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13648826 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31792979 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13979687 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13648839 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14031998 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793021 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793042 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31793045 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31792978 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10456416 - -
Affymetrix Mouse Gene 1.0 ST Array 10456420 - -
Affymetrix Mouse Gene 1.0 ST Array 10456419 - -
Affymetrix Mouse Gene 1.0 ST Array 10456415 - -
Affymetrix Mouse Gene 1.0 ST Array 10456421 - -
Affymetrix Mouse Gene 1.0 ST Array 10456417 - -
Affymetrix Mouse Gene 1.0 ST Array 10456422 - -
Affymetrix Mouse Gene 1.0 ST Array 10456418 - -
Affymetrix HT MG-430 PM Array Plate 1448212_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1444264_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10456414 - -
Mouse Genome 430 2.0 Array 1444264_PM_at - -
Mouse Genome 430 2.0 Array 1425373_PM_a_at - -
Mouse Genome 430 2.0 Array 1448212_PM_at - -
Mouse Genome 430 2.0 Array 1425373_a_at - -
Mouse Genome 430 2.0 Array 1444264_at - -
Mouse Genome 430 2.0 Array 1448212_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 107047_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 107047_PM_at - -
Affymetrix Mouse Clariom S Array PSR1800005187.mm.1 NM_134138.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005187.mm.1 NM_134138.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005187.mm.1 TC1800000708.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800005186.mm.1 NM_134138.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005186.mm.1 NM_134138.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005186.mm.1 TC1800000708.mm.1 MTA TC
Mouse Expression Set 430 1425373_PM_a_at - -
Mouse Expression Set 430 1448212_PM_at - -
Mouse Expression Set 430 1425373_a_at - -
Mouse Expression Set 430 1448212_at - -
Murine Genome U74v2 Set 99129_at - -
Murine Genome U74v2 Set 99129_PM_at - -
Mouse Genome 430A 2.0 Array 1448212_PM_at - -
Mouse Genome 430A 2.0 Array 1448212_at - -
Mouse Genome 430A 2.0 Array 1425373_PM_a_at - -
Mouse Genome 430A 2.0 Array 1425373_a_at - -
Affymetrix HT Mouse Genome 430A Array 1448212_at - -
Affymetrix HT Mouse Genome 430A Array 1448212_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 850381 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 2060095 - -
Illumina mouse-6 v1.0 expression beadchip 2060095 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002060095 - -
mouse-6 v1.1 (Illumina) scl51823.8.1_71-S - -
Illumina Sentrix mouse-6.v1 bead array 2060095 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl51823.8.1_71-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2060095 - -
MouseRef-8 v2 Expression BeadChip ILMN_3153450 - -
MouseRef-8 v2 Expression BeadChip ILMN_3074516 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3153450 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3074516 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1224609 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl51823.8.1_71-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-xbtwq:80/100.66.78.227:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0