This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00519796_g1
See other PLA2G16 GE Assays ›
Gene Symbol:
Pla2g16
Gene Name:
phospholipase A2, group XVI
Gene Aliases:
C78643, HRSL3, Hrasls3, Hrev107, MLP-3
Chromosome Location:
Chr.19: 7557434 - 7588545 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00519796_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pla2g16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_139269.2 NP_644675.2 1 - 2 102 86
GenBank mRNA AK042007.1 - 1 - 2 55 86
AK052657.1 - 1 - 2 81 86
AK163505.1 - 1 - 2 103 86
BC024581.1 - 1 - 2 48 5065247 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000141.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK042007.1,AK052657.1,AK163505.1,BC024581.1,NM_139269.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pla2g16

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
phospholipid metabolic process
phospholipid biosynthetic process
lipid catabolic process
negative regulation of cell cycle
phospholipase A2 activity
phosphatidylcholine 1-acylhydrolase activity
transferase activity, transferring acyl groups
hydrolase activity
phosphatidylserine 1-acylhydrolase activity
1-acyl-2-lysophosphatidylserine acylhydrolase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900001824.mm.1 NM_139269.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900001824.mm.1 NM_139269.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900001824.mm.1 TC1900000141.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900001825.mm.1 NM_139269.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900001825.mm.1 NM_139269.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900001825.mm.1 TC1900000141.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900001828.mm.1 NM_139269.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900001828.mm.1 NM_139269.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900001828.mm.1 TC1900000141.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 844982 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0