This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00519848_m1
See other IL31RA GE Assays ›
Gene Symbol:
Il31ra
Gene Name:
interleukin 31 receptor A
Gene Aliases:
CRL3, GLM-R, GPL, Glmr, NR10
Chromosome Location:
Chr.13: 112521849 - 112591399 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00519848_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il31ra
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_139299.2 NP_647460.2 15 - 16 2022 91
GenBank mRNA AB083111.1 - 15 - 16 2021 91
AF486621.1 - 12 - 13 1601 91
AK030512.1 - 4 - 5 591 91
AY509150.1 - 1918 91
BC145083.1 - 15 - 16 2031 91
BC145084.1 - 14 - 15 1925 91
BC145085.1 - 15 - 16 2048 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002679.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Il31ra

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

immune system process
positive regulation of cell proliferation
cytokine-mediated signaling pathway
cytokine receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300018118.mm.1 NM_139299.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300018118.mm.1 NM_139299.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300018118.mm.1 TC1300002679.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0