This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00520218_m1
See other NSUN2 GE Assays ›
Gene Symbol:
Nsun2
Gene Name:
NOL1/NOP2/Sun domain family member 2
Gene Aliases:
D13Wsu123e, Misu
Chromosome Location:
Chr.13: 69612016 - 69635780 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00520218_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nsun2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145354.5 NP_663329.3 6 - 7 698 95
GenBank mRNA AK030124.1 - 5 - 6 583 95
AK171399.1 - 6 - 7 654 95
BC013625.1 - 2 - 3 170 4011674 95
BC025549.1 - 6 - 7 662 95
DQ490066.1 - 6 - 7 661 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002766.mm.1

Additional Information:

This assay is in the 5'UTR region of the BC013625.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nsun2

Panther Classification:

Molecular Function -

RNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
tRNA processing
tRNA methylation
methylation
cell division
tRNA binding
RNA binding
methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity
transferase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300005494.mm.1 NM_145354.5 Refseq
Affymetrix Mouse Clariom S Array PSR1300005494.mm.1 TC1300002766.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0