This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00520276_m1
See other CREB3L3 GE Assays ›
Gene Symbol:
Creb3l3
Gene Name:
cAMP responsive element binding protein 3-like 3
Gene Aliases:
BC010786, CREB-H, D10Bur1e
Chromosome Location:
Chr.10: 81084324 - 81098872 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00520276_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Creb3l3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145365.3 NP_663340.1 5 - 6 876 68
GenBank mRNA AF392874.1 - 3 - 4 527 68
AK050186.1 - 3 - 4 518 68
AK077258.1 - 5 - 6 877 68
BC010786.1 - 3 - 4 571 4211480 68
BC028820.1 - 3 - 4 548 5096773 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002525.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Creb3l3

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
response to unfolded protein
endoplasmic reticulum unfolded protein response
positive regulation of transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
transcription regulatory region sequence-specific DNA binding
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
cAMP response element binding
protein homodimerization activity
sequence-specific DNA binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000018873.mm.1 NM_145365.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000018873.mm.1 NM_145365.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000018873.mm.1 TC1000002525.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-bbcf2:80/100.66.78.33:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline