This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00520419_m1
See other SLC44A3 GE Assays ›
Gene Symbol:
Slc44a3
Gene Name:
solute carrier family 44, member 3
Gene Aliases:
BC010552
Chromosome Location:
Chr.3: 121459528 - 121532344 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00520419_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc44a3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145394.3 NP_663369.2 4 - 5 568 110
GenBank mRNA AK145169.1 - 2 - 3 180 110
BC010552.1 - 3 - 4 336 3582543 110
BC025548.1 - 4 - 5 566 5320841 110
BY735109.1 - 2 - 3 180 110
CJ130557.1 - 1 - 2 69 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002840.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc44a3

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300022508.mm.1 NM_145394.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300022508.mm.1 NM_145394.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300022508.mm.1 TC0300002840.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-94gr6:80/100.66.78.133:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0