This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00520484_m1
See other TMPRSS4 GE Assays ›
Gene Symbol:
Tmprss4
Gene Name:
transmembrane protease, serine 4
Gene Aliases:
mCAP2
Chromosome Location:
Chr.9: 45172726 - 45204075 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00520484_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmprss4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145403.2 NP_663378.1 9 - 10 1189 70
GenBank mRNA AK078890.1 - 9 - 10 1100 70
AY043240.1 - 9 - 10 1189 70
BC021368.1 - 9 - 10 1138 5030266 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002196.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmprss4

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity
scavenger receptor activity
peptidase activity
serine-type peptidase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900018297.mm.1 NM_145403.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900018297.mm.1 NM_145403.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900018297.mm.1 TC0900002196.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900018298.mm.1 NM_145403.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900018298.mm.1 NM_145403.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900018298.mm.1 TC0900002196.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline