This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00520662_m1
See other ATPAF2 GE Assays ›
Gene Symbol:
Atpaf2
Gene Name:
ATP synthase mitochondrial F1 complex assembly factor 2
Gene Aliases:
ATP12, ATP12p
Chromosome Location:
Chr.11: 60400624 - 60418791 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00520662_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atpaf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145427.2 NP_663402.2 5 - 6 720 65
GenBank mRNA AK078500.1 - 5 - 6 665 65
BC013607.1 - 5 - 6 626 3585582 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002874.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atpaf2

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

biological_process
proton-transporting ATP synthase complex assembly
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27842892 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27842886 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27842891 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27842890 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27842888 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059650 BC013607.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059650 NM_145427.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059650 AK078500.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P438701 BC013607.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P438701 BE991226.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P438701 CJ303030.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P438701 NM_145427.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P438701 BC013607.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P438701 BE991226.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P438701 CJ303030.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P438701 NM_145427.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847445 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597747 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 740201 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 301970 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 894406 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 880996 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 220128 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 371890 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549100 AK035998.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549100 AK038855.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549100 AK158459.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549100 NM_001033394.2 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89069 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292171 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 97302 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86684 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100625 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161026 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P438701 BC013607.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P438701 BE991226.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P438701 CJ303030.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P438701 NM_145427.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P438701 BC013607.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P438701 BE991226.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P438701 CJ303030.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P438701 NM_145427.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059650 BC013607.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059650 NM_145427.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059650 AK078500.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P438701 BC013607.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P438701 BE991226.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P438701 CJ303030.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P438701 NM_145427.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P438701 BC013607.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P438701 BE991226.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P438701 CJ303030.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P438701 NM_145427.2 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27842871 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27842879 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27842873 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27842870 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27842881 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27842874 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27842890 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27842875 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27842868 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27842866 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27842892 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27842883 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27842882 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27842876 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27842869 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27842885 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27842888 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161026 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02339185 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100625 - -
Whole Mouse Genome G4122A A_52_P336268 BC013607.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P336268 NM_145427.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P336268 AK078500.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P438701 BC013607.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P438701 BE991226.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P438701 CJ303030.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P438701 NM_145427.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P438701 BC013607.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P438701 BE991226.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P438701 CJ303030.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P438701 NM_145427.2 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132706 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114223 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161026 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100625 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09424588 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09424583 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09508579 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27842886 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27842890 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27842891 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27842888 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27842892 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842873 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842812 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842810 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842856 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842888 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842829 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842861 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842828 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842892 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842804 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842843 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842826 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842863 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842857 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842823 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842830 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842866 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842822 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842793 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842832 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842874 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842879 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842842 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842820 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842807 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842816 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842781 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842801 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842885 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842853 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842871 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842854 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842827 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842850 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842881 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842836 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842795 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842802 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842809 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842890 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842852 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842814 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842839 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842876 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842825 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842848 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842875 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842800 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842835 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842798 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27842870 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842866 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842780 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842794 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842854 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09519363 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842798 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09351008 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842876 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842805 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842856 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842879 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842885 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842865 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842881 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842804 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09302407 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842779 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842888 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842883 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842850 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842873 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842870 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842861 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842868 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842801 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09686485 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842853 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842781 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842871 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842795 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842802 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09628025 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842875 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842874 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842852 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842857 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09017300 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842800 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842783 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27842858 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10386521 - -
Affymetrix Mouse Gene 1.0 ST Array 10386525 - -
Affymetrix Mouse Gene 1.0 ST Array 10386520 - -
Affymetrix Mouse Gene 1.0 ST Array 10386523 - -
Affymetrix Mouse Gene 1.0 ST Array 10386526 - -
Affymetrix Mouse Gene 1.0 ST Array 10386519 - -
Affymetrix Mouse Gene 1.0 ST Array 10386524 - -
Affymetrix Mouse Gene 1.0 ST Array 10386522 - -
Affymetrix HT MG-430 PM Array Plate 1426474_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10386518 - -
Mouse Genome 430 2.0 Array 1426474_at - -
Mouse Genome 430 2.0 Array 1426474_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 246782_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 246782_PM_at - -
Affymetrix Mouse Clariom S Array PSR1100026137.mm.1 NM_145427.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026137.mm.1 NM_145427.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026137.mm.1 TC1100002874.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026138.mm.1 NM_145427.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026138.mm.1 NM_145427.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026138.mm.1 TC1100002874.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026136.mm.1 NM_145427.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026136.mm.1 NM_145427.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026136.mm.1 TC1100002874.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026139.mm.1 NM_145427.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026139.mm.1 NM_145427.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026139.mm.1 TC1100002874.mm.1 MTA TC
Mouse Expression Set 430 1426474_at - -
Mouse Expression Set 430 1426474_PM_at - -
Mouse Genome 430A 2.0 Array 1426474_PM_at - -
Mouse Genome 430A 2.0 Array 1426474_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 349385 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 103170309 - -
Illumina mouse-6 v1.0 expression beadchip 6290750 - -
Illumina mouse-6 v1.0 expression beadchip 103170309 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006290750 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2601118 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1235311 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2601118 - -
mouse-6 v1.1 (Illumina) GI_21703851-S - -
mouse-6 v1.1 (Illumina) scl40146.10.1_50-S - -
Illumina Sentrix mouse-6.v1 bead array 103170309 - -
Illumina Sentrix mouse-6.v1 bead array 6290750 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl40146.10.1_50-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 103170309 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6290750 - -
MouseRef-8 v2 Expression BeadChip ILMN_2601118 - -
MouseRef-8 v2 Expression BeadChip ILMN_2959285 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2959285 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2601118 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40146.10.1_50-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0