This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00521936_g1
See other LIG3 GE Assays ›
Gene Symbol:
Lig3
Gene Name:
ligase III, DNA, ATP-dependent
Gene Aliases:
D11Wsu78e
Chromosome Location:
Chr.11: 82781109 - 82804274 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00521936_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lig3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291245.1 NP_001278174.1 7 - 8 1466 81
NM_001291246.1 NP_001278175.1 7 - 8 1463 81
NM_001291247.1 NP_001278176.1 7 - 8 1467 81
NM_010716.3 NP_034846.2 7 - 8 1470 81
GenBank mRNA AK031540.1 - 7 - 8 1407 81
AK143531.1 - 7 - 8 1336 81
AK150644.1 - 7 - 8 1375 81
AK156018.1 - 7 - 8 1385 81
BC026143.1 - 7 - 8 1438 81
BC049240.1 - 7 - 8 1373 4504384 81
BC083500.1 - 7 - 8 1369 3673103 81
U66057.1 - 7 - 8 1401 81
U66058.1 - 7 - 8 1401 81

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lig3

Panther Classification:

Molecular Function -

DNA ligase

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
DNA ligation
DNA repair
base-excision repair, DNA ligation
double-strand break repair
double-strand break repair via nonhomologous end joining
DNA recombination
cellular response to DNA damage stimulus
mitochondrion organization
cell cycle
mitochondrial DNA repair
negative regulation of DNA recombination
DNA ligation involved in DNA repair
cell division
DNA biosynthetic process
negative regulation of mitochondrial DNA replication
double-strand break repair via alternative nonhomologous end joining
nucleotide binding
DNA binding
DNA ligase activity
DNA ligase (ATP) activity
protein binding
ATP binding
zinc ion binding
ligase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0