This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00521972_m1
See other MBD4 GE Assays ›
Gene Symbol:
Mbd4
Gene Name:
methyl-CpG binding domain protein 4
Gene Aliases:
Med1
Chromosome Location:
Chr.6: 115840697 - 115853438 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00521972_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mbd4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010774.2 NP_034904.2 3 - 4 1256 117
GenBank mRNA AF072249.1 - 3 - 4 1273 117
BC024812.1 - 3 - 4 1238 5356016 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002963.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mbd4

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
base-excision repair
DNA methylation
cellular response to DNA damage stimulus
mitotic G2 DNA damage checkpoint
intrinsic apoptotic signaling pathway in response to DNA damage
response to radiation
purine-specific mismatch base pair DNA N-glycosylase activity
DNA binding
catalytic activity
uracil DNA N-glycosylase activity
pyrimidine-specific mismatch base pair DNA N-glycosylase activity
hydrolase activity
single-strand selective uracil DNA N-glycosylase activity
DNA N-glycosylase activity
DNA-3-methylbase glycosylase activity
nicotinamide riboside hydrolase activity
nicotinic acid riboside hydrolase activity
deoxyribonucleoside 5'-monophosphate N-glycosidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600022774.mm.1 NM_010774.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600022774.mm.1 NM_010774.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600022774.mm.1 TC0600002963.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline