This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00522086_g1
See other MADCAM1 GE Assays ›
Gene Symbol:
Madcam1
Gene Name:
mucosal vascular addressin cell adhesion molecule 1
Gene Aliases:
AV211525
Chromosome Location:
Chr.10: 79664395 - 79668541 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00522086_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Madcam1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013591.2 NP_038619.2 2 - 3 359 84
GenBank mRNA AK137028.1 - 2 - 3 360 84
AK156096.1 - 2 - 3 337 84
BC024372.1 - 2 - 3 352 4986802 84
BC115910.1 - 2 - 3 353 40105157 84
BC118056.1 - 2 - 3 353 40105156 84
L21203.1 - 2 - 3 356 84
S80258.1 - 2 - 3 326 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000808.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Madcam1

Gene Ontology Categories:

Function(s)

positive regulation of leukocyte migration
cell adhesion
keratinocyte differentiation
leukocyte migration

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 852164 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0