This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00522331_m1
See other BCAM GE Assays ›
Gene Symbol:
Bcam
Gene Name:
basal cell adhesion molecule
Gene Aliases:
1200005K12Rik, B-CAM, Gplu, Lu
Chromosome Location:
Chr.7: 19756138 - 19770532 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00522331_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bcam
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020486.2 NP_065232.1 1 - 2 124 95
GenBank mRNA AB035511.1 - 1 - 2 85 95
AF109160.1 - 1 - 2 124 95
AF221507.1 - 1 - 2 74 95
AF346663.1 - 1 - 2 62 95
AK040564.1 - 1 - 2 108 95
AK054001.1 - 1 - 2 105 95
AK075704.1 - 1 - 2 89 95
AK158439.1 - 1 - 2 84 95
BC004826.1 - 1 - 2 108 2616148 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002500.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bcam

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
cell-matrix adhesion
laminin receptor activity
protein C-terminus binding
laminin binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700022117.mm.1 TC0700002500.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700022117.mm.1 NM_020486.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700022117.mm.1 NM_020486.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700022119.mm.1 TC0700002500.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700022119.mm.1 NM_020486.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700022119.mm.1 NM_020486.2 Refseq
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 363825 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0