This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00522337_g1
See other BCAM GE Assays ›
Gene Symbol:
Bcam
Gene Name:
basal cell adhesion molecule
Gene Aliases:
1200005K12Rik, B-CAM, Gplu, Lu
Chromosome Location:
Chr.7: 19756138 - 19770532 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00522337_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bcam
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020486.2 NP_065232.1 7 - 8 962 67
GenBank mRNA AB035511.1 - 7 - 8 923 67
AF109160.1 - 7 - 8 962 67
AF221507.1 - 7 - 8 912 67
AF346663.1 - 7 - 8 900 67
AK040564.1 - 7 - 8 946 67
AK054001.1 - 7 - 8 943 67
AK075704.1 - 7 - 8 927 67
BC004826.1 - 7 - 8 946 2616148 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002500.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bcam

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
cell-matrix adhesion
laminin receptor activity
protein C-terminus binding
laminin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700022110.mm.1 TC0700002500.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700022110.mm.1 NM_020486.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700022110.mm.1 NM_020486.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0