This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00522470_m1
See other LIAS GE Assays ›
Gene Symbol:
Lias
Gene Name:
lipoic acid synthetase
Gene Aliases:
2900022L22Rik, 4933425M12Rik, 7a5ex, C77512, mLip1
Chromosome Location:
Chr.5: 65391497 - 65409208 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00522470_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lias
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024471.4 NP_077791.1 3 - 4 437 109
GenBank mRNA AB057731.1 - 3 - 4 336 109
AK031851.1 - 3 - 4 409 109
AK045167.1 - 3 - 4 222 109
AK048744.1 - 381 109
AK049548.1 - 3 - 4 352 109
AK077192.1 - 3 - 4 438 109
BC002141.1 - 3 - 4 369 3484559 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000611.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lias

Gene Ontology Categories:

Function(s) Process(es)

neural tube closure
inflammatory response
response to oxidative stress
lipoate biosynthetic process
response to lipopolysaccharide
catalytic activity
transferase activity
lipoate synthase activity
metal ion binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500005444.mm.1 TC0500000611.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500005444.mm.1 NM_024471.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0