This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00522673_g1
See other PYCR1 GE Assays ›
Gene Symbol:
Pycr1
Gene Name:
pyrroline-5-carboxylate reductase 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 120635712 - 120643782 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00522673_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pycr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144795.3 NP_659044.1 1 - 2 511 77
GenBank mRNA AK139185.1 - 1 - 2 512 77
BC006727.1 - 2 - 3 156 3962093 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100004222.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pycr1

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

proline biosynthetic process
cellular amino acid biosynthetic process
cellular response to oxidative stress
regulation of mitochondrial membrane potential
oxidation-reduction process
negative regulation of hydrogen peroxide-induced cell death
pyrroline-5-carboxylate reductase activity
oxidoreductase activity
identical protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100040384.mm.1 NM_144795.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100040384.mm.1 NM_144795.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100040384.mm.1 TC1100004222.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100040385.mm.1 NM_144795.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100040385.mm.1 NM_144795.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100040385.mm.1 TC1100004222.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100040388.mm.1 NM_144795.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100040388.mm.1 NM_144795.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100040388.mm.1 TC1100004222.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100040387.mm.1 NM_144795.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100040387.mm.1 NM_144795.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100040387.mm.1 TC1100004222.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100040386.mm.1 NM_144795.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100040386.mm.1 NM_144795.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100040386.mm.1 TC1100004222.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 560198 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0