This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00522986_g1
See other SLC7A4 GE Assays ›
Gene Symbol:
Slc7a4
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 4
Gene Aliases:
AI853530
Chromosome Location:
Chr.16: 17572018 - 17576892 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00522986_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144852.3 NP_659101.2 3 - 4 1687 69
GenBank mRNA AK043772.1 - 3 - 4 1688 69
AK162398.1 - 3 - 4 1731 69
BC016100.1 - 4 - 5 1305 4911158 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001294.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc7a4

Panther Classification:

Molecular Function -

amino acid transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
amino acid transport
biological_process
L-arginine import across plasma membrane
L-ornithine transmembrane transport
L-lysine transmembrane transport
L-ornithine transmembrane transporter activity
molecular_function
amino acid transmembrane transporter activity
arginine transmembrane transporter activity
L-lysine transmembrane transporter activity
antiporter activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30574555 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30574556 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30574558 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30574557 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999775 AK043772.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999775 NM_144852.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1997400 AK162398.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1997400 AK030586.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1997400 AK043772.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1997400 NM_144852.3 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 706382 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 927188 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105681 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999775 AK043772.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999775 NM_144852.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1997400 AK162398.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1997400 AK030586.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1997400 AK043772.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1997400 NM_144852.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P311597 AK162398.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P311597 BC016100.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P311597 AK030586.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P311597 AK043772.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P311597 NM_144852.3 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574535 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574549 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574550 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574571 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574564 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574553 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574534 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574576 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574537 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574569 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574542 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574555 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574547 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574531 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574562 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574541 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574574 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574543 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574561 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574567 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574546 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574548 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574573 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574559 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574558 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574539 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30574532 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105681 - -
Whole Mouse Genome G4122A A_52_P311597 AK162398.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P311597 BC016100.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P311597 AK030586.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P311597 AK043772.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P311597 NM_144852.3 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101998 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138484 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105681 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12591580 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12591544 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12591579 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12591548 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12591552 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12591564 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12591553 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12591546 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30574557 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30574556 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30574555 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30574558 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574546 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574550 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574532 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574585 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574449 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574467 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574517 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574488 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574589 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574524 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574535 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574476 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574571 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574469 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574461 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574487 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574481 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574441 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574579 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574458 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574526 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574447 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574542 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574520 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574534 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574537 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574561 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574558 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574486 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574573 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574559 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574569 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574512 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574479 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574472 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574530 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574539 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574528 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574483 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574555 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574583 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574519 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574444 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574457 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574473 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574475 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574548 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574564 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574464 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574439 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574470 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574514 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574553 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574574 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574463 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574567 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574453 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574455 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574521 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574460 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574515 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574442 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574543 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574580 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574521 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574577 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12736527 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574532 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574547 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574569 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574539 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574555 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574561 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12736515 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12736521 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574546 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574567 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574580 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574541 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574531 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574528 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12591583 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574530 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574559 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574524 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574520 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574579 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12521163 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12591536 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574534 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574517 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574543 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12591543 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574514 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12591563 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12591551 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574576 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574574 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12591534 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12591564 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574537 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12591562 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30574558 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12591553 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12693740 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10438193 - -
Affymetrix Mouse Gene 1.0 ST Array 10438196 - -
Affymetrix Mouse Gene 1.0 ST Array 10438194 - -
Affymetrix Mouse Gene 1.0 ST Array 10438192 - -
Affymetrix Mouse Gene 1.0 ST Array 10438190 - -
Affymetrix Mouse Gene 1.0 ST Array 10438195 - -
Affymetrix Mouse Gene 1.0 ST Array 10438191 - -
Affymetrix Mouse Gene 1.0 ST Array 10438197 - -
Affymetrix HT MG-430 PM Array Plate 1426068_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1436776_PM_x_at - -
Affymetrix HT MG-430 PM Array Plate 1426069_PM_s_at - -
Affymetrix Mouse Gene 1.1 ST Array 10438189 - -
Mouse Genome 430 2.0 Array 1426068_PM_at - -
Mouse Genome 430 2.0 Array 1436776_x_at - -
Mouse Genome 430 2.0 Array 1426068_at - -
Mouse Genome 430 2.0 Array 1436776_PM_x_at - -
Mouse Genome 430 2.0 Array 1426069_PM_s_at - -
Mouse Genome 430 2.0 Array 1426069_s_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 224022_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 224022_at - -
Affymetrix Mouse Clariom S Array PSR1600010529.mm.1 NM_144852.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010529.mm.1 NM_144852.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600010529.mm.1 TC1600001294.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600010530.mm.1 NM_144852.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010530.mm.1 NM_144852.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600010530.mm.1 TC1600001294.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600010526.mm.1 NM_144852.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010526.mm.1 NM_144852.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600010526.mm.1 TC1600001294.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600010527.mm.1 NM_144852.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010527.mm.1 NM_144852.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600010527.mm.1 TC1600001294.mm.1 MTA TC
Mouse Expression Set 430 1426068_at - -
Mouse Expression Set 430 1426068_PM_at - -
Mouse Expression Set 430 1426069_s_at - -
Mouse Expression Set 430 1426069_PM_s_at - -
Mouse Expression Set 430 1436776_x_at - -
Mouse Expression Set 430 1436776_PM_x_at - -
Mouse Genome 430A 2.0 Array 1426068_at - -
Mouse Genome 430A 2.0 Array 1436776_x_at - -
Mouse Genome 430A 2.0 Array 1426069_s_at - -
Mouse Genome 430A 2.0 Array 1426068_PM_at - -
Mouse Genome 430A 2.0 Array 1436776_PM_x_at - -
Mouse Genome 430A 2.0 Array 1426069_PM_s_at - -
Affymetrix HT Mouse Genome 430A Array 1436776_x_at - -
Affymetrix HT Mouse Genome 430A Array 1426069_PM_s_at - -
Affymetrix HT Mouse Genome 430A Array 1426068_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1426069_s_at - -
Affymetrix HT Mouse Genome 430A Array 1436776_PM_x_at - -
Affymetrix HT Mouse Genome 430A Array 1426068_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 333040 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 106770168 - -
MouseWG-6 v2 Expression BeadChip 5080711 - -
MouseWG-6 v2 Expression BeadChip 1190168 - -
Illumina mouse-6 v1.0 expression beadchip 106770168 - -
Illumina mouse-6 v1.0 expression beadchip 1190168 - -
Illumina mouse-6 v1.0 expression beadchip 5080711 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001190168 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005080711 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2639360 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2640131 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1252581 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2640131 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2639360 - -
mouse-6 v1.1 (Illumina) scl0001800.1_60-S - -
mouse-6 v1.1 (Illumina) scl48704.9_493-S - -
mouse-6 v1.1 (Illumina) scl00224022.2_173-S - -
Illumina Sentrix mouse-6.v1 bead array 5080711 - -
Illumina Sentrix mouse-6.v1 bead array 106770168 - -
Illumina Sentrix mouse-6.v1 bead array 1190168 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl00224022.2_173-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0001800.1_60-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1190168 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5080711 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 106770168 - -
MouseRef-8 v2 Expression BeadChip ILMN_2639360 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2639360 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1252581 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00224022.2_173-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0