This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00523203_m1
See other SLC35C2 GE Assays ›
Gene Symbol:
Slc35c2
Gene Name:
solute carrier family 35, member C2
Gene Aliases:
C85957, CGI-15, D2Wsu58e, Ovcov1
Chromosome Location:
Chr.2: 165276522 - 165287888 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00523203_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc35c2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252573.1 NP_001239502.1 8 - 9 1179 97
NM_001252574.1 NP_001239503.1 8 - 9 1114 97
NM_001252575.1 NP_001239504.1 8 - 9 1062 97
NM_144893.2 NP_659142.1 8 - 9 1133 97
NR_045544.1 9 - 10 1010 97
GenBank mRNA AK053180.1 - 8 - 9 1049 97
AK155539.1 - 8 - 9 1148 97
AK160899.1 - 8 - 9 1119 97
AK170715.1 - 8 - 9 1084 97
AK195585.1 - 1 - 2 44 97
BC018327.1 - 9 - 10 999 4166249 97
BC094025.1 - 8 - 9 1112 4488690 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200005145.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc35c2

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
negative regulation of gene expression
UDP-glucose transport
fucosylation
protein O-linked fucosylation
positive regulation of Notch signaling pathway
anion transmembrane transport
molecular_function
antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200043897.mm.1 NM_001252573.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200043897.mm.1 NM_001252573.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200043897.mm.1 NM_001252574.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200043897.mm.1 NM_001252574.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200043897.mm.1 NM_001252575.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200043897.mm.1 NM_001252575.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200043897.mm.1 NM_144893.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200043897.mm.1 NM_144893.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200043897.mm.1 NR_045544.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200043897.mm.1 NR_045544.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200043897.mm.1 TC0200005145.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0