This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00523297_mH
See other ALDOB GE Assays ›
Gene Symbol:
Aldob
Gene Name:
aldolase B, fructose-bisphosphate
Gene Aliases:
Aldo-2, Aldo2, BC016435
Chromosome Location:
Chr.4: 49535995 - 49549483 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00523297_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aldob
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144903.2 NP_659152.1 6 - 7 686 85
GenBank mRNA AK135385.1 - 6 - 7 715 85
AK149395.1 - 6 - 7 703 85
AK149598.1 - 6 - 7 700 85
AK167334.1 - 6 - 7 703 85
AK167566.1 - 6 - 7 700 85
AK168600.1 - 6 - 7 700 85
AK168997.1 - 6 - 7 750 85
BC016435.1 - 6 - 7 682 4241025 85
BC022113.1 - 6 - 7 659 5098524 85
BC024056.1 - 6 - 7 692 5100067 85
BC024112.1 - 6 - 7 691 5102118 85
BC026577.1 - 6 - 7 651 4989521 85
BC030724.1 - 6 - 7 688 5100784 85
BC030725.1 - 6 - 7 692 5101686 85
BC034169.1 - 6 - 7 692 4975382 85
BC034171.1 - 6 - 7 669 4972521 85
BC034172.1 - 6 - 7 656 4974489 85
BC034173.1 - 6 - 7 668 4974553 85
BC036130.1 - 6 - 7 669 4974071 85
BC036131.1 - 6 - 7 686 4974877 85
BC036132.1 - 6 - 7 686 4975336 85
BC036133.1 - 6 - 7 698 4975573 85
BC036139.1 - 2 - 3 310 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002638.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aldob

Panther Classification:

Molecular Function -

aldolase

Gene Ontology Categories:

Function(s) Process(es)

fructose metabolic process
glycolytic process
NADH oxidation
response to zinc ion
fructose 1,6-bisphosphate metabolic process
positive regulation of ATPase activity
response to peptide hormone
vacuolar proton-transporting V-type ATPase complex assembly
catalytic activity
fructose-bisphosphate aldolase activity
cytoskeletal protein binding
lyase activity
phosphatidylcholine binding
identical protein binding
ATPase binding
fructose-1-phosphate aldolase activity
fructose binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400021489.mm.1 TC0400002638.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400021489.mm.1 NM_144903.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400021488.mm.1 TC0400002638.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400021488.mm.1 NM_144903.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0