This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00523540_m1
See other UROC1 GE Assays ›
Gene Symbol:
Uroc1
Gene Name:
urocanase domain containing 1
Gene Aliases:
AI265605, BC022133
Chromosome Location:
Chr.6: 90333239 - 90364551 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00523540_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uroc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144940.2 NP_659189.2 17 - 18 1733 81
GenBank mRNA AK149464.1 - 17 - 18 1836 81
BC022133.1 - 17 - 18 1716 5098655 81
BC033269.1 - 14 - 15 1348 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001071.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Uroc1

Panther Classification:

Molecular Function -

hydratase lyase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

histidine metabolic process
histidine catabolic process
urocanate hydratase activity
lyase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600008301.mm.1 TC0600001071.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600008301.mm.1 NM_144940.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600008301.mm.1 NM_144940.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0