This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00523701_g1
See other P2RX3 GE Assays ›
Gene Symbol:
P2rx3
Gene Name:
purinergic receptor P2X, ligand-gated ion channel, 3
Gene Aliases:
4930513E20Rik, P2X3
Chromosome Location:
Chr.2: 84996396 - 85037029 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00523701_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2rx3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145526.2 NP_663501.2 3 - 4 869 109
GenBank mRNA AK140283.1 - 3 - 4 2137 109
AK141847.1 - 3 - 4 534 109
BC023089.1 - 3 - 4 522 3984739 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003885.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene P2rx3

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
transport
ion transport
cation transport
chemical synaptic transmission
neuromuscular synaptic transmission
response to temperature stimulus
response to heat
response to cold
response to mechanical stimulus
response to carbohydrate
response to organic substance
urinary bladder smooth muscle contraction
neuronal action potential
peristalsis
response to ATP
purinergic nucleotide receptor signaling pathway
regulation of synaptic plasticity
behavioral response to pain
sensory perception of taste
protein homooligomerization
cation transmembrane transport
purinergic nucleotide receptor activity
extracellular ATP-gated cation channel activity
ion channel activity
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200033628.mm.1 NM_145526.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200033628.mm.1 NM_145526.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200033628.mm.1 TC0200003885.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200033626.mm.1 NM_145526.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200033626.mm.1 NM_145526.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200033626.mm.1 TC0200003885.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-c7pcq:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0