This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00524570_g1
See other ARHGAP9 GE Assays ›
Gene Symbol:
Arhgap9
Gene Name:
Rho GTPase activating protein 9
Gene Aliases:
AU043488
Chromosome Location:
Chr.10: 127321959 - 127329946 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00524570_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285785.1 NP_001272714.1 7 - 8 1100 120
NM_146011.2 NP_666123.2 6 - 7 539 120
GenBank mRNA AK047280.1 - 6 - 7 484 120
AK049245.1 - 3 - 4 320 120
AK080955.1 - 5 - 6 1234 120
AK083474.1 - 3 - 4 449 120
AK089051.1 - 6 - 7 491 120
AK167738.1 - 7 - 8 677 120
BC024535.1 - 6 - 7 534 4987970 120
BC027374.1 - 6 - 7 510 4947306 120
DQ498128.1 - 7 - 8 948 120

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000001560.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK047280.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap9

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of GTPase activity
GTPase activator activity
phosphatidylinositol-3,4,5-trisphosphate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000011118.mm.1 NM_001285785.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000011118.mm.1 NM_146011.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000011118.mm.1 NM_146011.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000011118.mm.1 TC1000001560.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0