This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00524572_g1
See other ARHGAP9 GE Assays ›
Gene Symbol:
Arhgap9
Gene Name:
Rho GTPase activating protein 9
Gene Aliases:
AU043488
Chromosome Location:
Chr.10: 127321959 - 127329946 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00524572_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285785.1 NP_001272714.1 9 - 10 1341 81
NM_146011.2 NP_666123.2 8 - 9 780 81
GenBank mRNA AK047280.1 - 8 - 9 725 81
AK049245.1 - 5 - 6 561 81
AK080955.1 - 6 - 7 1696 81
AK083474.1 - 5 - 6 690 81
AK089051.1 - 8 - 9 732 81
BC024535.1 - 8 - 9 775 4987970 81
BC027374.1 - 8 - 9 751 4947306 81
DQ498128.1 - 9 - 10 1189 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000001560.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK047280.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap9

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of GTPase activity
GTPase activator activity
phosphatidylinositol-3,4,5-trisphosphate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000011120.mm.1 NM_001285785.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000011120.mm.1 NM_146011.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000011120.mm.1 NM_146011.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000011120.mm.1 TC1000001560.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rzthw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0