Assay Details
| Gene Symbol: Arhgap9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285785.1 | NP_001272714.1 | 11 - 12 | 1555 | 62 | |
| NM_146011.2 | NP_666123.2 | 10 - 11 | 994 | 62 | ||
| GenBank mRNA | AK047280.1 | - | 10 - 11 | 939 | 62 | |
| AK049245.1 | - | 7 - 8 | 775 | 62 | ||
| AK080955.1 | - | 8 - 9 | 1910 | 62 | ||
| AK083474.1 | - | 7 - 8 | 904 | 62 | ||
| AK089051.1 | - | 10 - 11 | 946 | 62 | ||
| AK167738.1 | - | 11 - 12 | 1132 | 62 | ||
| BC024535.1 | - | 10 - 11 | 989 | 4987970 | 62 | |
| BC027374.1 | - | 10 - 11 | 965 | 4947306 | 62 | |
| DQ498128.1 | - | 11 - 12 | 1403 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap9
Genomic Map