This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00524932_g1
See other APBB3 GE Assays ›
Gene Symbol:
Apbb3
Gene Name:
amyloid beta (A4) precursor protein-binding, family B, member 3
Gene Aliases:
Fe65l2, Rirl2, TR2S
Chromosome Location:
Chr.18: 36671154 - 36679366 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00524932_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apbb3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146085.1 NP_666197.1 10 - 11 1229 103
GenBank mRNA AK141878.1 - 10 - 11 1131 103
AK171634.1 - 9 - 10 1422 103
BC024136.1 - 9 - 10 1227 103
BC024809.1 - 10 - 11 1229 5357681 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001748.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Apbb3

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
negative regulation of apoptotic process
beta-amyloid binding
transcription factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800008807.mm.1 NM_146085.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800008807.mm.1 NM_146085.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800008807.mm.1 TC1800001748.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0