Assay Details
| Gene Symbol: Nrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146150.2 | NP_666262.2 | 13 - 14 | 1676 | 127 | |
| GenBank mRNA | AK031548.1 | - | 13 - 14 | 1676 | 127 | |
| AK040377.1 | - | 12 - 13 | 1907 | 127 | ||
| BC023786.1 | - | 12 - 13 | 1054 | 127 | ||
| BC026832.1 | - | 13 - 14 | 1726 | 4486176 | 127 | |
| BC029977.1 | - | 4 - 5 | 312 | 127 | ||
| BC036128.1 | - | 13 - 14 | 1794 | 4971791 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nrd1
Genomic Map