Assay Details
| Gene Symbol: Olfr1346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146916.1 | NP_667127.1 | 1 | 651 | 114 | |
| GenBank mRNA | BC137876.1 | - | 1 | 675 | 8860895 | 114 |
| BC137877.1 | - | 1 | 675 | 8860896 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Olfr1346
Genomic Map