This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00545815_m1
See other AGER GE Assays ›
Gene Symbol:
Ager
Gene Name:
advanced glycosylation end product-specific receptor
Gene Aliases:
RAGE
Chromosome Location:
Chr.17: 34597410 - 34600937 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00545815_m1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ager
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271422.1 NP_001258351.1 7 - 8 863 108
NM_001271423.1 NP_001258352.1 7 - 8 836 108
NM_001271424.1 NP_001258353.1 6 - 7 958 108
NM_007425.3 NP_031451.2 7 - 8 863 108
NR_073173.1 7 - 8 863 108
NR_073175.1 6 - 7 1058 108
NR_073176.1 6 - 7 1058 108
GenBank mRNA AB207883.1 - 7 - 8 826 108
AK004802.1 - 7 - 8 853 108
BC061182.1 - 7 - 8 851 30300427 108
EU520325.1 - 7 - 8 819 108
EU570240.1 - 7 - 8 819 108
EU570241.1 - 6 - 7 1014 108
EU570242.1 - 7 - 8 819 108
EU570243.1 - 7 - 8 819 108
EU570244.1 - 6 - 7 1014 108
EU570245.1 - 6 - 7 1014 108
EU570246.1 - 6 - 7 976 108
EU570247.1 - 6 - 7 1014 108
EU906863.1 - 7 - 8 792 108
EU906864.1 - 6 - 7 1014 108
GU164727.1 - 6 - 7 933 108
GU164730.1 - 7 - 8 838 108
GU164733.1 - 2 - 3 188 108
KC692918.1 - 7 - 8 819 108
L33412.1 - 7 - 8 827 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000637.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ager

Panther Classification:

Molecular Function -

cell adhesion molecule immunoglobulin superfamily cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
negative regulation of endothelial cell proliferation
inflammatory response
negative regulation of cell adhesion
JAK-STAT cascade
positive regulation of autophagy
negative regulation of endothelial cell migration
positive regulation of gene expression
positive regulation of epithelial to mesenchymal transition
positive regulation of fibroblast migration
positive regulation of smooth muscle cell migration
positive regulation of cell migration
neuron projection development
negative regulation of collagen biosynthetic process
positive regulation of apoptotic process
positive regulation of JUN kinase activity
positive regulation of neuron apoptotic process
positive regulation of fibroblast proliferation
positive regulation of smooth muscle cell proliferation
regulation of inflammatory response
positive regulation of inflammatory response
induction of positive chemotaxis
positive regulation of NF-kappaB transcription factor activity
regulation of DNA binding
calcium ion homeostasis
positive regulation of phagocytosis, engulfment
protein localization to membrane
positive regulation of potassium ion transmembrane transporter activity
positive regulation of neuron death
positive regulation of endothelial cell apoptotic process
positive regulation of reactive oxygen species metabolic process
positive regulation of type B pancreatic cell apoptotic process
protein binding
identical protein binding
S100 protein binding
advanced glycation end-product receptor activity
high mobility group box 1 binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 558424 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0