Assay Details
| Gene Symbol: Tssk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009435.2 | NP_033461.2 | 1 | 1347 | 68 | |
| GenBank mRNA | AK006517.1 | - | 1 | 1347 | 68 | |
| AK029655.1 | - | 1329 | 68 | |||
| AK076822.1 | - | 1 | 1323 | 68 | ||
| BC050772.1 | - | 1 | 1320 | 6772455 | 68 | |
| U01840.1 | - | 1 | 1251 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
multicellular organism development
spermatogenesis
spermatid development
phosphorylation
cell differentiation
intracellular signal transduction
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
metal ion binding
protein phosphorylation
multicellular organism development
spermatogenesis
spermatid development
phosphorylation
cell differentiation
intracellular signal transduction
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426347 | BC050772.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426347 | U01840.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426347 | AK006517.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426347 | AK029655.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426347 | AK076822.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426347 | BC050772.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426347 | U01840.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426347 | AK006517.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426347 | AK029655.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426347 | AK076822.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 7739 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 959151 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426347 | BC050772.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426347 | U01840.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426347 | AK006517.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426347 | AK029655.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426347 | AK076822.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426347 | BC050772.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426347 | U01840.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426347 | AK006517.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426347 | AK029655.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426347 | AK076822.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576373 | - | - |
| Whole Mouse Genome G4122A | A_52_P665682 | AK029655.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426347 | BC050772.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426347 | U01840.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426347 | AK006517.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426347 | AK029655.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426347 | AK076822.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426347 | BC050772.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426347 | U01840.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426347 | AK006517.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426347 | AK029655.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426347 | AK076822.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122248 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12541115 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12541121 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12541098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576375 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576377 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12541115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12541071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12541070 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10434122 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417989_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10434121 | - | - |
| Mouse Genome 430 2.0 Array | 1417989_at | - | - |
| Mouse Genome 430 2.0 Array | 1417989_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22114_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22114_PM_at | - | - |
| Mouse Expression Set 430 | 1417989_PM_at | - | - |
| Mouse Expression Set 430 | 1417989_at | - | - |
| Murine Genome U74v2 Set | 103360_at | - | - |
| Murine Genome U74v2 Set | 103360_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417989_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417989_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417989_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417989_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 104850575 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4480537 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480537 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2762918 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251399 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2762918 | - | - |
| mouse-6 v1.1 (Illumina) | scl0022114.2_229-S | - | - |
| mouse-6 v1.1 (Illumina) | scl49368.2_121-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480537 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104850575 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0022114.2_229-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480537 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104850575 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1251399 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1251399 | - | - |

Tssk1
Genomic Map