Assay Details
| Gene Symbol: Rnf121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029211.2 | NP_083487.2 | 7 - 8 | 758 | 66 | |
| GenBank mRNA | AK016037.1 | - | 7 - 8 | 989 | 66 | |
| AK034806.1 | - | 7 - 8 | 2072 | 66 | ||
| AK078509.1 | - | 7 - 8 | 719 | 66 | ||
| AK159032.1 | - | 7 - 8 | 895 | 66 | ||
| AK159260.1 | - | 7 - 8 | 785 | 66 | ||
| AK168821.1 | - | 7 - 8 | 771 | 66 | ||
| AK169089.1 | - | 7 - 8 | 771 | 66 | ||
| BC022686.1 | - | 7 - 8 | 758 | 4241984 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089293 | AK168821.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089293 | AK169089.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089293 | BC022686.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089293 | AK078509.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089293 | AK159032.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093226 | AK168821.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093226 | AK169089.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093226 | BC022686.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093226 | AK078509.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093226 | AK159032.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089288 | AK168821.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089288 | AK169089.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089288 | AK159032.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089288 | AK159260.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 881656 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 430133 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 956697 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489930 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830802 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 180724 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 806734 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 533640 | AK044313.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 131111 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 195085 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 667018 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 613467 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 474281 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741735 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 662883 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 651887 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 946035 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 89251 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 627844 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593069 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 210502 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 320055 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 682233 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 98638 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 833067 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 709767 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59616 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 797093 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13680 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 33960 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 110903 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52781 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 37101 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093226 | AK168821.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093226 | AK169089.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093226 | BC022686.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093226 | AK078509.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093226 | AK159032.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089293 | AK168821.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089293 | AK169089.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089293 | BC022686.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089293 | AK078509.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089293 | AK159032.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089288 | AK168821.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089288 | AK169089.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089288 | AK159032.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089288 | AK159260.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P189234 | AK168821.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P189234 | AK169089.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P189234 | BC022686.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P189234 | AK078509.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P189234 | AK159032.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P408802 | AK168821.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P408802 | AK169089.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P408802 | AK078509.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P408802 | AK159032.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P408802 | AK159260.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P117482 | AK016037.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137319 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06668381 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05987574 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06668377 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05987567 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05987579 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06668384 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05987571 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10566073 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566070 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566076 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566077 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566071 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566074 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566072 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566069 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566075 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426504_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426503_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10566067 | - | - |
| Mouse Genome 430 2.0 Array | 1426504_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1426503_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1426503_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1426504_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75212_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75212_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700032522.mm.1 | NM_029211.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032522.mm.1 | NM_029211.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032522.mm.1 | TC0700003851.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700032523.mm.1 | NM_029211.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032523.mm.1 | NM_029211.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032523.mm.1 | TC0700003851.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700032521.mm.1 | NM_029211.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032521.mm.1 | NM_029211.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032521.mm.1 | TC0700003851.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700032524.mm.1 | NM_029211.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032524.mm.1 | NM_029211.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032524.mm.1 | TC0700003851.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426503_PM_a_at | - | - |
| Mouse Expression Set 430 | 1426504_PM_a_at | - | - |
| Mouse Expression Set 430 | 1426504_a_at | - | - |
| Mouse Expression Set 430 | 1426503_a_at | - | - |
| Murine Genome U74v2 Set | 168287_i_at | - | - |
| Murine Genome U74v2 Set | 168287_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426504_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426503_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426503_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426504_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426504_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426503_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426504_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426503_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104480152 | - | - |
| MouseWG-6 v2 Expression BeadChip | 110079 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104780091 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 110079 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104480152 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000110079 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161576 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223096 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223096 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161576 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2422239 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2558964 | - | - |
| mouse-6 v1.1 (Illumina) | GI_70778931-S | - | - |
| mouse-6 v1.1 (Illumina) | scl30951.12_249-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0075212.2_206-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 110079 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104480152 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104780091 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0075212.2_206-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104780091 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104480152 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 110079 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2903529 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223096 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2903529 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2558964 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2422239 | - | - |

Rnf121
Genomic Map