This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00547077_g1
See other ARHGAP12 GE Assays ›
Gene Symbol:
Arhgap12
Gene Name:
Rho GTPase activating protein 12
Gene Aliases:
2810011M08Rik
Chromosome Location:
Chr.18: 6024448 - 6136098 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00547077_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039692.1 NP_001034781.1 17 - 18 2649 130
NM_029277.2 NP_083553.2 16 - 17 2574 130
GenBank mRNA AK031648.1 - 17 - 18 2493 130
AK077063.1 - 17 - 18 2650 130
BC024633.1 - 12 - 13 1061 130
BC037608.1 - 16 - 17 2390 131
BC141184.1 - 16 - 17 2383 9055796 130
BC145506.1 - 16 - 17 2382 9054069 130

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000944.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap12

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

morphogenesis of an epithelial sheet
signal transduction
molecular_function
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800006883.mm.1 NM_001039692.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800006883.mm.1 NM_001039692.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800006883.mm.1 NM_029277.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800006883.mm.1 NM_029277.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800006883.mm.1 TC1800000944.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0