This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00549588_g1
See other CCBL1 GE Assays ›
Gene Symbol:
Ccbl1
Gene Name:
cysteine conjugate-beta lyase 1
Gene Aliases:
2010009K05Rik, AI182306, Gtk, Kat1, KatI
Chromosome Location:
Chr.2: 30185124 - 30206295 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00549588_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ccbl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172404.2 NP_765992.2 8 - 9 964 60
GenBank mRNA AK031511.1 - 8 - 9 977 60
AK042391.1 - 7 - 8 910 60
AK088404.1 - 7 - 8 872 60
AK163165.1 - 7 - 8 772 60
AK207151.1 - 2 - 3 208 60
AK218327.1 - 2 - 3 208 60
BC016206.1 - 8 - 9 1066 4912335 60
BC025124.1 - 7 - 8 1097 4486058 60
BC052047.1 - 8 - 9 964 5687475 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003265.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ccbl1

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

pyruvate metabolic process
biosynthetic process
kynurenine metabolic process
L-kynurenine metabolic process
catalytic activity
transaminase activity
kynurenine-oxoglutarate transaminase activity
transferase activity
lyase activity
pyridoxal phosphate binding
protein homodimerization activity
L-phenylalanine:pyruvate aminotransferase activity
glutamine-phenylpyruvate transaminase activity
cysteine-S-conjugate beta-lyase activity
L-glutamine:pyruvate aminotransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200027581.mm.1 NM_172404.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027581.mm.1 NM_172404.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200027581.mm.1 TC0200003265.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline