This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00549718_m1
See other ASXL2 GE Assays ›
Gene Symbol:
Asxl2
Gene Name:
additional sex combs like 2 (Drosophila)
Gene Aliases:
4930556B16Rik, mKIAA1685
Chromosome Location:
Chr.12: 3426857 - 3506852 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00549718_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Asxl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001270988.1 NP_001257917.1 10 - 11 1455 86
NM_172421.5 NP_766009.2 9 - 10 1311 86
GenBank mRNA AK016141.1 - 9 - 10 1017 86
AK035679.1 - 4 - 5 1883 86
AK036839.1 - 9 - 10 1268 86
AK080679.1 - 10 - 11 1429 86
AK122540.1 - 1 - 2 84 86
AK148015.1 - 9 - 10 1316 86
AK162455.1 - 1 - 2 52 86
BC141140.1 - 10 - 11 1289 9055752 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200000009.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Asxl2

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
heart morphogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
heart development
adult heart development
positive regulation of peroxisome proliferator activated receptor signaling pathway
positive regulation of fat cell differentiation
positive regulation of osteoclast differentiation
positive regulation of transcription from RNA polymerase II promoter
embryonic skeletal system morphogenesis
positive regulation of bone mineralization involved in bone maturation
positive regulation of histone H3-K27 trimethylation
DNA binding
peroxisome proliferator activated receptor binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200000104.mm.1 NM_001270988.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000104.mm.1 NM_172421.5 Refseq
Affymetrix Mouse Clariom S Array PSR1200000104.mm.1 TC1200000009.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200000106.mm.1 NM_001270988.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000106.mm.1 NM_172421.5 Refseq
Affymetrix Mouse Clariom S Array PSR1200000106.mm.1 TC1200000009.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200000105.mm.1 NM_001270988.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000105.mm.1 NM_172421.5 Refseq
Affymetrix Mouse Clariom S Array PSR1200000105.mm.1 TC1200000009.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0