This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00549805_g1
See other CELF3 GE Assays ›
Gene Symbol:
Celf3
Gene Name:
CUGBP, Elav-like family member 3
Gene Aliases:
4930415M08Rik, BRUNOL1, CAGH4, ERDA4, Tnrc4
Chromosome Location:
Chr.3: 94478285 - 94492199 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00549805_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Celf3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289616.1 NP_001276545.1 9 - 10 961 110
NM_172434.3 NP_766022.1 8 - 9 1476 110
GenBank mRNA AK019578.1 - 5 - 6 1387 110
AK044506.1 - 9 - 10 925 110
AY165052.1 - 8 - 9 1194 110

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Celf3

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
mRNA processing
spermatogenesis
RNA splicing
sperm motility
positive regulation of mRNA splicing, via spliceosome
nucleotide binding
nucleic acid binding
RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline