This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00549875_m1
See other PIF1 GE Assays ›
Gene Symbol:
Pif1
Gene Name:
PIF1 5'-to-3' DNA helicase homolog (S. cerevisiae)
Gene Aliases:
4631410M14, AI449441
Chromosome Location:
Chr.9: 65587147 - 65595967 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00549875_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pif1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172453.3 NP_766041.1 1 - 2 1192 75
GenBank mRNA AK028460.1 - 1 - 2 1193 75
AK049353.1 - 1 - 2 1194 75
AK154072.1 - 1 - 2 1205 75
AY100322.1 - 2 - 3 905 75
AY498715.1 - 1 - 2 1192 75
BC046611.1 - 2 - 3 830 6396273 75
BC060108.1 - 1 - 2 1183 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000847.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pif1

Panther Classification:

Molecular Function -

DNA helicase

Gene Ontology Categories:

Function(s) Process(es)

telomere maintenance
DNA repair
DNA recombination
cellular response to DNA damage stimulus
regulation of telomere maintenance
DNA duplex unwinding
negative regulation of telomerase activity
nucleotide binding
magnesium ion binding
DNA binding
DNA helicase activity
helicase activity
ATP binding
hydrolase activity
single-stranded DNA-dependent ATP-dependent DNA helicase activity
ATP-dependent 5'-3' DNA/RNA helicase activity
telomeric DNA binding
ATP-dependent 5'-3' DNA helicase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900006471.mm.1 NM_172453.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006471.mm.1 NM_172453.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006471.mm.1 TC0900000847.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900006470.mm.1 NM_172453.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006470.mm.1 NM_172453.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006470.mm.1 TC0900000847.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900006475.mm.1 NM_172453.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006475.mm.1 NM_172453.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006475.mm.1 TC0900000847.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900006473.mm.1 NM_172453.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006473.mm.1 NM_172453.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006473.mm.1 TC0900000847.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900006474.mm.1 NM_172453.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006474.mm.1 NM_172453.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006474.mm.1 TC0900000847.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900006468.mm.1 NM_172453.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006468.mm.1 NM_172453.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006468.mm.1 TC0900000847.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900006469.mm.1 NM_172453.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006469.mm.1 NM_172453.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006469.mm.1 TC0900000847.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900006472.mm.1 NM_172453.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006472.mm.1 NM_172453.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006472.mm.1 TC0900000847.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 832418 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0